Print…
Orf clones, ANGPTL6 cDNA, Homo sapiens
Fornecedor: Genscript
GENSNM_031917EA 467 EUR
GENSNM_031917
Orf clones, ANGPTL6 cDNA, Homo sapiens
Vectors, Plasmids, Libraries and Inserts
ANGPTL6 gene cDNA ORF clone sequences were retrieved from the NCBI Reference Sequence Database (RefSeq). These sequences represent the protein coding region of the ANGPTL6 cDNA ORF which is encoded by the open reading frame (ORF) sequence.
RefSeq accession NM_031917.2: The customised vector for this sequence is pcDNA3,1+/C-(K)DYK.