Print…

Anti-CD39 Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB319728EA 0 EUR
BIRBORB319728
Anti-CD39 Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CD39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG
Reactivity: Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR