Print…

Anti-NKp46 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB157934-100EA 0 EUR
BIRBORB157934-100
Anti-NKp46 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: NKp46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR