Print…

Anti-KLRG2 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB184535-100EA 0 EUR
BIRBORB184535-100
Anti-KLRG2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: KLRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR