Print…
Laboratory syringe pump, KDS 210
Fornecedor: Ismatec
ISMCKDS210EA 5810 EUR
ISMCKDS210
Laboratory syringe pump, KDS 210
The KDS 210 including five operating modes plus independent rate and volume settings for both infusion and withdrawal.
  • Vibration elimination system
  • Very constant flow rate in the nanoliter range
  • Convey and dose pulsation
  • High chemical resistance
Multiple mode selection such as infusion, withdrawl, infusion and withdrawl, withdrawl and infusion and continuous cycle.
Certificações: CE Approved