Print…
Urine containers, Eco friendly SpecTainer™
Fornecedor: Simport
SIMPC567-60AQSECOEA 89.4 EUR
SIMPC567-60AQSECO SIMPC566-90DOECO SIMPC566-120DOECO SIMPC566-60DOECO SIMPC566-90AQSECO SIMPC566-120AQSECO SIMPC567-60DOECO SIMPC567-120AQSECO SIMPC566-60AQSECO SIMPC567-90AQSECO SIMPC567-120DOECO SIMPC567-90DOECO
Urine containers, Eco friendly SpecTainer™
Sistemas para diagnóstico Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP urine container with tamper evident or conventional aqua (sterile) or gold (non sterile) PE screw cap, ideal for collecting, storing and transporting urine samples.
  • Does not contain heavy metals
  • No negative effect to its physical properties
  • No toxic residue
  • Fully degradable within seven years, aerobically or anaerobically
  • Label has space for patient identification
The Eco-Friendly SpecTainer™ have the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment. Does not require heat, physical stress and sunlight for the degradation.