Print…

Anti-CD6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornecedor: Bioss
BOSSBS-2488R-HRPEA 858 EUR
BOSSBS-2488R-HRP
Anti-CD6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpos
Involved in cell adhesion. Binds to CD166.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CD6
Clonality: Polyclonal
Clone: 0
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR