Print…
Chemical resistant cape hood, model 516, MICROCHEM® 4000
Fornecedor: Ansell
Microchem® 4000
ANSEGR40T-00516-00EA 979 EUR
ANSEGR40T-00516-00
Chemical resistant cape hood, model 516, MICROCHEM® 4000
Roupas Descartáveis
This cape hood with visor is made from a multi-layer barrier fabric (external film barrier, non woven inner layer, core high barrier film) that is renowned for its lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals.
  • Double zip system helps ensure a liquid tight seal without the need for additional taping
  • Hood with heat sealed visor, rear vent and 3 inch velcro body panel
  • Ultrasonically welded and taped seams
  • Textile-like inner improves wearer acceptance
  • Permeation tested against over 180 chemicals, including chemical warfare agents
Typical applications include: Chemical handling/transportation, oil-based mud protection, hazardous waste remediation, sewage purification installations, industrial/tank cleaning, HAZMAT Emergency Response (i.e. Level B), pharmaceutical, mining, agriculture.
Certificações: Type 3/4/5, EN 14126, EN 1073-2, EN 1149-5