Print…
Microcentrifuge tubes, 1,5 ml, disposable
VWRI525-0990EA 0 EUR
VWRI525-0990
Microcentrifuge tubes, 1,5 ml, disposable
PP, non sterile, autoclavable, graduated, flat caps and frosted areas on the sides for easy labelling. Amber coloured for light-sensitive substances.
  • Snap caps can be penetrated by a syringe
  • Easy to open caps will not pop open during centrifuging
  • Certified nuclease- (RNase-/DNase-), endotoxin- (pyrogen-) and ATP-free
Embalagem: Packed in resealable bags.