Print…

Anti-SMARCAD1 Rabbit Polyclonal Antibody

Fornecedor: Bioworld Technology
BWRLBS7898EA 341 EUR
BWRLBS7898
Anti-SMARCAD1 Rabbit Polyclonal Antibody
Anticorpos
Recombinant full length Human SMARCAD1.

SMARCAD1 pAb detects endogenous levels of SMARCAD1 protein.

Type: Primary
Antigen: SMARCAD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR