Print…

Anti-CYP2A6 Rabbit Polyclonal Antibody [clone: E131]

Fornecedor: Bioworld Technology
BWRLBS2193EA 262 EUR
BWRLBS2193
Anti-CYP2A6 Rabbit Polyclonal Antibody [clone: E131]
Anticorpos
Synthetic peptide, corresponding to amino acids 101-150 of Human CYP2A6.

CYP2A6 (E131) pAb detects endogenous levels of CYP2A6 protein.

Type: Primary
Antigen: CYP2A6
Clonality: Polyclonal
Clone: E131
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR