Print…
Automated nucleic acid purification system, Maxwell® RSC instrument
PROMAS4500EA 31468.5 EUR
PROMAS4500
Automated nucleic acid purification system, Maxwell® RSC instrument
The Maxwell® Rapid Sample Concentrator (RSC) instrument is a platform for automated purification of nucleic acid from a range of sample types.
  • Up to 16 samples can be prepared simultaneously in 25 – 60 minutes
  • High quality nucleic acid purification with minimal steps and less hands-on time
  • Intuitive software and touch screen interface