Print…
Safety funnels with level control
Fornecedor: SCAT Europe
SCTV117637EA 288 EUR
SCTV117637
Safety funnels with level control
Made of electrostatic conductive HDPE. Space saving construction, suitable for every safety cabinet.
  • Lightweight construction
  • Integrated level control
  • No more overflowing containers
  • Grounding cable and clamp included
  • Removable sieve for easy cleaning
Keep filling levels in non-transparent containers safe and easy under control.