Print…
Cleanroom benches, Micro CRB and Pharma CRB models, TINMAN
Fornecedor: Tinman
TINMMB2000350450EA 1026 EUR
TINMMB2000350450 TINMMB600350450 TINMMB1800350450 TINMPB1000350450 TINMMB800350450 TINMMB1000350450 TINMMB1500350450 TINMMB1200350450
Cleanroom benches, Micro CRB and Pharma CRB models, TINMAN
Benches designed to assist in putting on shoe covers, are ideal in all gowning areas. CRB benches can be used in many different ways, providing comfort in gowning operation.
  • Ergonomic design: rounded front and back edge – higher comfort level
  • Smooth edges prevents tearing and cuts
  • Adjustable feet: non-slip leveling feet, anchored leveling feet
  • With perforations: no retention of cleaning fluids; air permeability 40% (only Micro Micro CRB models)
  • Minisation of gaps: closed circuit welded joints; preventing accumulation of dust
  • Good clean ability due to low surface roughness and easy cleaning access
  • Very good resistance against chemicals and all regular cleanroom disinfectants (alcoholic, biocidal and sporicidal)
  • Simple assembly
Two different models are available, the Micro CRB model with a perforated bench top and the Pharma CRB model a smooth bench top with its good clean ability due to low surface roughness and easy cleaning access.
Material: AISI 304, 2R - high gloss polished (optional AISI 316)
Cleanroom suitability: ISO class 4, GMP class A
Embalagem: Flat Detailed assembly instructions in every package.