Print…
Pocket TDS tester, TDSTestr 1™
Fornecedor: MP Biomedicals
ICNA0215948301EA 198 EUR
ICNA0215948301
Pocket TDS tester, TDSTestr 1™
The MP TDSTestr 1™ offers convenient and rapid measurement of Total Dissolved Solids.
  • Large, easy-to-read LCD with mode indicator
  • Full Scale readout (in ppm or ppt), eliminating the need for multiplying the displayed reading to obtain the actual test values
  • Automatic temperature compensation (ATC)
  • ±2% full scale accuracy
  • HOLD function
Fornecimento: Supplied with four 1,4 V batteries.