Print…
Centrifuges, ventilated/refrigerated, floor-standing, ROTIXA 50 S / 50 RS
Fornecedor: Hettich
523-4272EA 0 EUR
HETT4910 HETT4905
Centrifuges, ventilated/refrigerated, floor-standing, ROTIXA 50 S / 50 RS
Centrifuges of choice in development and quality control laboratories of the life science industry and in medical routine laboratories.
  • Smooth and quiet running, exceptional speed stability
  • Frequency-controlled drive, maintenance only required according to UVV regulations, nine acceleration/braking ramps with extremely short run-up and run-down times
  • 89 programmable memories
  • Safety features: Panelling with scratch and impact resistant coating, automatic lid locking and holding, lid dropping protection, emergency lid lock release, automatic rotor recognition, imbalance switch-off, key-operated switch for locking and protecting programs, motor overheating protection, chamber overheating protection for ROTIXA 50 RS
  • Options: Centrifuging data report system HettInfo (for easy data documentation); Barcode reader; Visual indication when the rotor is stationary; Chaining of programs (allowing to program the centrifuge to run different programs in succession); Second independent tacho system for calibrating the speed measuring instrument as required by quality management systems
Suited for separating a wide range of samples such as food and beverage, cell suspensions, sediments of chemical reactions, sludge and, of course, blood and blood components.
Informações de encomenda: Rotors and adapters available separately.