Print…
HIV-1 p24 ELISA kit
Fornecedor: Abnova
ABNOKA0452EA 878 EUR
ABNOKA0452
HIV-1 p24 ELISA kit
The HIV-1 p24 ELISA Kit is a colorimetric, sandwich enzyme immunoassay for the quantitative determination of HIV-1 p24 in culture supernatants and tissue.
  • Calibration range: 12,5 to 200 pg/ml
  • Limit of detection: 1,7 pg/ml
  • Sample volume: 200 µl
Fornecimento: The kit contains coated microwell srip 96(8×12) wells, positive p24 control 10 ng/ml (0,1 ml), lysis buffer (6 ml), detector(12 ml), conjugate (12 ml), wash buffer (60 ml×2), substrate solution (12 ml) and stop solution (12 ml).