The Clino® CR4 EM-CR trolley system made of stainless steel suitable for cleanrooms and for high sensitive areas that do not require sterility.
- Individually configurable through Clino modular design system
- EasyMop CR with ergonomically optimised system box
- Highest efficiency and process reliability in single step technique
- EasyMop system boxes available in red, green, blue, yeollow, grey and athracite
- Includes contact-free dropping device MopDrop
- Cloth impregnation for surface cleaning in PlusBox
- Basic module mini with high end finish and castors (100 mm)
A small cleaning system with pre-preparation system EasyMop for technical cleanrooms suitable for ISO classes 7 and 8 or GMP areas C/D and it allows a validatable and reproducible results.
Fornecimento: Supplied with 1×tray for Clino Systembox, 2×system boxes EasyMop CR, 2×PlusBoxes, disposal unit (1×35 L) and plastic container (5 L).