Print…
Temperature and humidity data loggers, SATO SK-L200TH II
SIMEHM8020EA 532 EUR
SIMEHM8020
Temperature and humidity data loggers, SATO SK-L200TH II
Extremely compact datalogger for recording temperature and air humidity.
  • Memory capacity is 8 100 lines of data per channel
  • Date and time to start logging automatically can be specified by setting the preset start
  • Measurement data can be transmitted in realtime to a personal computer
  • USB transmission is adopted
  • Upper and lower thresholds for warning can be set. When the temperature or humidity
  • threshold is triggered, the alarm mark (Hi or Lo) will light
  • Probes are interchangeably used with main units
  • Storage memory can be varified with the memory indicator
Log interval can be selected from 14 options: 1 second, 2 seconds, 5 seconds, 10 seconds, 15 seconds, 30 seconds, 1 minute, 2 minutes, 5 minutes, 10 minutes, 15 minutes, 30 minutes, 60 minutes and 90 minutes.
Informações de encomenda: Complete with attached combiprobe, USB cable and evaluation software. A wire-combiprobe is optional; contact your Avantor representative for more information.