Print…
Temperature loggers, CurveX-3 standard oven logger
Fornecedor: Industrial Physics
SIMECX3015EA 1710 EUR
SIMECX3015 SIMECX3020
Temperature loggers, CurveX-3 standard oven logger
CurveX 3 Standard is a 6-channel thermocouple K type input oven is used to monitoring the progress of the burning process of powder paint kilns.
  • Equipped with a large colour touch screen interface for easy operation
  • Operation via the menu on the display device
  • High-precision temperature logging
  • Measurement, analysis and reports are fully user-selectable
  • Compatible with ideal finish analysis software
  • Aluminium housing with protective sleeve
The data logger is fitted with a large full-color touchscreen for easy menu-driven operation and quick display of measurement results. CurveX 3 standard oven logger kit contains all USB cable, USB charger, USB stick with ideal finish software, probe ID kit, calibration certificate, manual CurveX 3 standard, manual ideal finish analysis, plastic carrying case, CX2005 insulation box, CX2011 heat absorber, just add the desired magnetic or clamp-type probes to make the oven logger KIT complete.
Fornecimento: Supplied with USB cable and charger, USB memory stick with Ideal Finish Analysis software, set of identification labels probes, factory calibrated calibration certificate and plastic case.