Print…
Disposable particle filtering respirators, 9310/9312
Fornecedor: 3M
111-0033EA 20 EUR
111-0033
Disposable particle filtering respirators, 9310/9312
Máscaras respiratórias Máscaras Descartáveis Máscaras respiratórias para partículas
The foldable disposable FFP1 respirators protect against dusts and mists. They are available with or without Cool Flow™ exhalation valve that helps to breathe easier when working for prolonged periods.
  • Unique 3-panel design fits a wide range of face shapes and sizes, accommodates facial movements and makes the respirator easy to talk through
  • Concealed adjustable nose-clip is metal detectable
  • Coloured straps for easy identification of protection class (yellow for P1)
  • Made from 3M™ Advanced Electret Filter Material for effective filtration with low breathing resistance
  • Do not contain components made from natural rubber latex
Protection factor: 10× Workplace Exposure Standard.
Certificações: AS/NZS 1716:2012