Print…
Procurou por: Reagentes para transfecção
80  resultados encontrados
SearchResultCount:"80"
Sort Results
Visualização em Lista Easy View (new)
Classifique os resultados da pesquisa
Fornecedor: Polyplus-transfection
Descrição: Next generation of transfection reagent for rAAV production in suspension.
Fornecedor: Polyplus-transfection
Descrição: Transfection reagents for lentiviral vector production from small to large scale.
Novo produto
Fornecedor: Polyplus-transfection
Descrição: The jetPRIME® DNA and/or siRNA transfection reagent is a powerful and versatile reagent designed to ensure high DNA transfection efficiency and excell...
Fornecedor: Polyplus-transfection
Descrição: jetOPTIMUS® is a 'best in class' DNA transfection reagent, based on a novel cationic nanotechnology allowing high efficiency with low volume of reagen...
Fornecedor: Polyplus-transfection
Descrição: in vivo-jetPEI® reagent provides versatile, reproducible and reliable delivery of all types of nucleic acids (DNA, siRNA, miRNA and more) in an...
Fornecedor: Polyplus-transfection
Descrição: jetMESSENGER® is a mRNA transfection reagent that has been specifically designed to offer outstanding transfection efficiency in cells that are usuall...
Fornecedor: Polyplus-transfection
Descrição: The FectoPRO® Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various se...
Código de Artigo: (AVIVGWB-300101)
Fornecedor: Aviva Systems Biology
Descrição: Reagente de transfecção, BaculoFECTIN II transfection reagent
UOM: 1 * 150 µl
Novo produto
Código de Artigo: (101000122.)
Fornecedor: Polyplus-transfection
Descrição: In vivo-jetRNA®+ is a ready-to-use, efficient and stable lipid-based transfection reagent optimised for in vivo mRNA delivery.
UOM: 1 * 1 mL
Código de Artigo: (722-100)
Fornecedor: Polyplus-transfection
Descrição: SpCas9 nuclease sequence is carefully optimized to improve its nuclear import, nuclease editing efficiency, and to reduce off-targeting effects
UOM: 1 * 100 µG
Código de Artigo: (201000002.)
Fornecedor: Polyplus-transfection
Descrição: This 150 mM NaCl solution is recommended as dilution reagent for DNA transfection with jetPEI®.
UOM: 1 * 50 mL
Código de Artigo: (201000005.)
Fornecedor: Polyplus-transfection
Descrição: Reagente de transfecção, Glucose solution
UOM: 1 * 10 mL
Fornecedor: Polyplus-transfection
Descrição: PEIpro® is a ready to use 1 mg/ml optimised linear polyethylenimine (PEI) reagent for the production of virus and recombinant proteins by Transient Ge...
Fornecedor: Polyplus-transfection
Descrição: FectoCHO® expression system consists in the synergistic association of a novel CHO chemically defined medium, FectoCHO® CD expression medium and a pow...
Fornecedor: Polyplus-transfection
Descrição: INTERFERin® is a powerful, ready to use siRNA and miRNA reagent which provides more than 90% silencing efficiency at 1 nM siRNA in a wide variety of c...
Fornecedor: Polyplus-transfection
Descrição: jetPEI® transfection reagent is a linear polyethylenimine derivative, free of components of animal origin, providing highly effective and reproducible...
Preço sob consulta
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call ainda é exibida e precisar de ajuda, por favor, ligue para 213 600 770
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call está visível e precisar de ajuda, por favor, ligue para 213 600 770
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão.
Este produto está bloqueado. Para obter mais informações, contacte a VWR através do número 213 600 770.
O produto pretendido já não se encontra disponível. O produto indicado é um substituto.
Este produto encontra-se em rutura de stock. Poderá encontrar alternativas pesquisando pelo código de artigo indicado acima. Se precisar de ajuda, por favor contacte a VWR através do 213 600 770.
1 - 16 of 80
no targeter for Bottom