Print…
Reusable half masks, Oxy K 30 HS
Fornecedor: Dräger
Oxy K 30 HW/HS
DRAE6304701EA 1260 EUR
DRAE6304701 DRAE6304700
Reusable half masks, Oxy K 30 HS
Máscaras respiratórias Máscaras Reutilizáveis Semi máscaras respiratórias
Oxy K 30 HS is an oxygen self-rescuer, independent of the ambient air by using chemically-bound oxygen. A quick starter cartridge provides an instant oxygen supply on donning.
  • Easy to don and breath
  • Easy maintenance and cleaning
  • It can be installed in vehicles or carried by users
Suitable for use in transportation, shipping, sewage, tunnelling, drilling platforms.