Print…
Centrifuge, benchtop, ventilated, SL 40
Fornecedor: Thermo Scientific
HERA75004512EA 5098 EUR
HERA75004512
Centrifuge, benchtop, ventilated, SL 40
The SL 40 centrifuge series offers exceptional capacity, ergonomic features and value for routine sample processing on the bench top. Advanced ergonomic design features proprietary innovations for safe, simple and comfortable daily operation.
  • Motorised lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches
  • AutoLock™–III rotor system: Secure locking system allows easy push-button installation and exchange of rotors
  • ClickSeal™ bucket sealing system: Biocontainment solution seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • SMARTSpin™ Technology: Advanced rotor management system maximises acceleration, braking and residual load imbalance for each rotor and bucket option
  • Up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing
Informações de encomenda: Supplied with power supply cable, instruction manual and anti-corrosion oil. Without rotors and adapters which need to be ordered separately.