Print…
Procurou por: Density Standards
16  resultados encontrados
SearchResultCount:"16"
Sort Results
Visualização em Lista Easy View (new)
Classifique os resultados da pesquisa
Fornecedor: Merck
Descrição: 2,2,4-Trimethylpentane, 692 kg/m³
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
Fornecedor: VWR Chemicals
Descrição: VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are flu...
Fornecedor: VWR Chemicals
Descrição: This range of relative density standards are certified in accordance with primary level ASTM D1480 methodology, and are dual-certified to ISO 17025 / ...
Fornecedor: VWR Chemicals
Descrição: VWR® Chemicals ISO 17025/ISO 17034 dual certified-density standards are for the verification of instruments used to measure the density of materials w...
Fornecedor: VWR Chemicals
Descrição: VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are flu...
Fornecedor: VWR Chemicals
Descrição: VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are flu...
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
Fornecedor: VWR Chemicals
Descrição: Density Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for density...
Código de Artigo: (85321.180)
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
UOM: 1 * 100 mL
MSDS Certificados
Código de Artigo: (85301.180)
Fornecedor: VWR Chemicals
Descrição: These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
UOM: 1 * 100 mL
MSDS Certificados
Fornecedor: BELLINGHAM & STANLEY
Descrição: Calibration materials for verifying the performance of digital density meters using the oscillating u-tube principle. Calibrated in accordance with I...
Novo produto
Código de Artigo: (614-1802)
Fornecedor: Mettler - Toledo
Descrição: Density standard, 0,998 g/cm³, certified at 20 °C, uncertainty (typically) 0,001 g/cm³, 6 ml sachets
UOM: 1 * 10 unid.
MSDS
Preço sob consulta
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call ainda é exibida e precisar de ajuda, por favor, ligue para 213 600 770
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call está visível e precisar de ajuda, por favor, ligue para 213 600 770
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão.
Este produto está bloqueado. Para obter mais informações, contacte a VWR através do número 213 600 770.
O produto pretendido já não se encontra disponível. O produto indicado é um substituto.
Este produto encontra-se em rutura de stock. Poderá encontrar alternativas pesquisando pelo código de artigo indicado acima. Se precisar de ajuda, por favor contacte a VWR através do 213 600 770.
1 - 16 of 16
no targeter for Bottom