Print…

Anti-SIX1 Rabbit Polyclonal Antibody

Fornecedor: Boster
BSBTPB9889EA 579 EUR
BSBTPB9889
Anti-SIX1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: SIX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR