Print…
Human recombinant GITRL
Fornecedor: Tonbo Biosciences
21-7146-U500EA 3140 EUR
TNBO21-7146-U500 TNBO21-7146-U005 TNBO21-7146-U100 TNBO21-7146-U020
Human recombinant GITRL
GITRL, also known as TNFSF18 and AITRL, along with its corresponding receptor, are members of the TNF superfamily. GITRL is expressed in endothelial cells and is thought to modulate T lymphocyte survival in peripheral tissues. It plays a role in interactions between activated T lymphocytes and endothelial cells. GITRL also triggers an increase in the capability of endothelial cells to support leukocyte adhesion.