Keep my session open?
Ending In 
The session is expired
A sua sessão expirou. Para sua segurança, fechámos a sua sessão.
Gostaria de reiniciar sessão?

Anti-IFN gamma Rabbit Polyclonal Antibody

Fornecedor: BioVision
IFN-gamma is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-gamma receptors are present in most immune cells, which respond to IFN-gamma signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-gamma signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-gamma stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Murine IFN-gamma is a 15.6 kDa protein containing 134 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Neutralization: 0.2-1 µg/ml

Type: Primary
Antigen: IFN gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse



Faça Login na página para visualizar o seu preço e disponibilidade dos artigos.
  Lembrar-me neste dispositivo
 

ENCOMENDA

Expandir tudo / Esconder tudo
Tamanho Und. Tipo de embalagem. Referência VWR Quantidade
100 µg 100 µG Vial BIOV5117-100
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF

ESPECIFICAÇÕES

Antigen symbol IFNG
Antigen name Interferon gamma
Reactivity Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms IFI, IFG
Gene ID 3458
Concentração 0,5 mg/ml
Cross adsorption No
Immunogen E. coli expressed murine IFN-gamma
Purification Affinity purified
Form Liquid
Storage buffer PBS, pH 7,2, containing 50% glycerol, and 0,02% thimerosal.
Temperatura de armazenamento – 20 °C
Shipping temperature 4 °C
Detalhes

Selecione o artigo para o qual procura o Certificado

Indique o número do lote. Este campo é obrigatório.

Inserir detalhes do certificado

Número de Lote:

Choose from recent batch/lot numbers:

Ou
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd