Print…

Anti-GM-CSF Rabbit Polyclonal Antibody

Fornecedor: BioVision
BIOV5100-100EA 276 EUR
BIOV5100-100
Anti-GM-CSF Rabbit Polyclonal Antibody
Human GM-CSF (Granulocyte Macrophage Colony Stimulating Factor) is a potent species-specific stimulator of bone marrow cells. GM-CSF stimulates precursor cells of granulocytes, macrophages and eosinophils.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR