Print…
Magnetic hotplate stirrers, SuperNuova™
Fornecedor: Thermo Scientific
BARNSP88857198EA 1300 EUR
BARNSP88857198 BARNSP88857195 BARNSP88850195 BARNSP131820-33Q BARNSP131820-33
Magnetic hotplate stirrers, SuperNuova™
Magnetic hotplates stirrers designed to provide reliable performance, dependable safety and simple operation. These units are available in two sizes and provide optimsed controls and settings for applications that demand advanced precision. Units features innovative engineering for slow-speed stirring, consistent speed control and strong magnetic coupling. Top plates are available in a choice of ceramic or aluminium.
  • Easy-to-read LED display for heating and stirring
  • Hot top warning indicator and automatic over temperature protection
  • Heating up time is 8 min to maximum temperature
Temperature uniformity (at 37 °C): ≤±2 °C