Print…
Sample concentrator, Chromacol™
Fornecedor: Thermo Scientific
CRMA60180-P990EA 12880 EUR
CRMA60180-P990 CRMA60180-P900
Sample concentrator, Chromacol™
This sample concentrator is designed for any SBS/ANSI 96 well plates.
  • Simple to install and operate
  • Quicker dry down times than standard methods such as vacuum oven
  • Easy adjustments of temperature, gas flow rates and needle depth into the wells