Print…
Earmuffs, Peltor™ WS™ Workstyle™
Fornecedor: 3M
Peltor™
MMMAWS5SVMEA 0 EUR
MMMAWS5SVM
Earmuffs, Peltor™ WS™ Workstyle™
This headset features Bluetooth® which allows wireless communication between the headset and mobile phones. The patented speaker system is specifically designed to produce a full-bodied, dynamic sound in an enclosed headset.
  • High attenuating hearing protector, allows you to listen to music and take phone calls via bluetooth®
  • Bluetooth® sound limiter and dosimeter, built in microphone
  • Level dependent function for ambient listening
  • Operating time approximately 12 hours full streaming; warning signal when power is low
  • Last settings stored when switched off
  • Stainless steel headband, foldable
SNR: 26 dB