Print…
Anti-SOAT-2/ACAT-2 Rabbit Polyclonal Antibody
CAYM100027-1EA 569 EUR
CAYM100027-1
Anti-SOAT-2/ACAT-2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: SOAT-2/ACAT-2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Sheep, Pig, Rat