Print…

Anti-ATG13 Rabbit Polyclonal Antibody

Fornecedor: Rockland Immunochemicals
ROCK600-401-C49EA 783 EUR
ROCK600-401-C49
Anti-ATG13 Rabbit Polyclonal Antibody
Anticorpos
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 56.6 kDa in size corresponding to human phosphorylated ATG13 Protein.

Recommended Dilutions: ELISA: 1:25000-1:175000; Western Blot: 1:1000;

Type: Primary
Antigen: ATG13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR