Print…

Anti-Collagen II Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBI139814EA 454 EUR
USBI139814
Anti-Collagen II Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Collagen II
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR