Print…

Anti-Zona Radiata Mouse Monoclonal Antibody [clone: 6D847]

Fornecedor: United States Biological
USBIZ0790-50EA 513 EUR
USBIZ0790-50
Anti-Zona Radiata Mouse Monoclonal Antibody [clone: 6D847]
Anticorpos
Recommended dilution: ELISA: 1:100-1:5000; Western Blot: 1:100-1:000

Type: Primary
Antigen: Zona Radiata
Clonality: monoclonal
Clone: 6D847
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR