Print…

Anti-TJP1 Rat Monoclonal Antibody [clone: 10H16]

Fornecedor: United States Biological
USBIZ0775GEA 729 EUR
USBIZ0775G
Anti-TJP1 Rat Monoclonal Antibody [clone: 10H16]
Anticorpos
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: TJP1 (tight junction protein 1)
Clonality: monoclonal
Clone: 10H16
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR