Print…

Anti-IKZF1 Goat Polyclonal Antibody

Fornecedor: United States Biological
USBIZ0749-05BEA 551 EUR
USBIZ0749-05B
Anti-IKZF1 Goat Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: IKZF1 (IKAROS family zinc finger 1 (Ikaros))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR