|
|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ZNF654 |
Antigen name | zinc finger protein 654 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | NKI/C3 |
Host | Mouse |
Isotype | IgG1 |
Formato | ascites |
Form | liquid |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |