|
|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ZBTB49 |
Antigen name | zinc finger and BTB domain containing 49 |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody type | Primary |
Clone | 8C375 |
Host | Mouse |
Isotype | IgG1 |
Western blot | Yes |
Concentração | ~0,1 mg/ml |
Purification | purified by Protein G chromatography |
Form | liquid |
Storage buffer | PBS, pH 7,4, 1% BSA, 0,05% sodium azide. |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |