Gostaria de reiniciar sessão?
Anti-ZNF496 Mouse monoclonal antibody unconjugated [clone: 11C983]
Fornecedor: US Biological
Recommended dilution: ELISA: 1 μg/ml-3ng/ml; Western Blot: 1:500-1:1000
Type: Primary
Antigen: ZNF496 (zinc finger protein 496)
Clonality: monoclonal
Clone: 11C983
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
ENCOMENDA
Expandir tudo / Esconder tudo
Tamanho Und. Referência VWR Quantidade
100 µl 100 µG USBIZ0744-96
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF
ESPECIFICAÇÕES
Antigen symbol ZNF496
Antigen name zinc finger protein 496
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 11C983
Host Mouse
Isotype IgG2a
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Amino acid number 485 to 586
Purification purified by immunoaffinity chromatography
Form liquid
Storage buffer PBS, pH 7,2.
Temperatura de armazenamento Cool
Shipping temperature Ice
Detalhes
Selecione o artigo para o qual procura o Certificado
Indique o número do lote. Este campo é obrigatório.
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping