|
|
ENCOMENDA
ESPECIFICAÇÕES
Antigen symbol | ZBTB2 |
Antigen name | zinc finger and BTB domain containing 2 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Epitope | C-terminal |
Purification | purified by ammonium sulfate precipitation |
Form | liquid |
Storage buffer | PBS, 0,09% sodium azide. |
Temperatura de armazenamento | Cool |
Shipping temperature | Ice |