Print…
Anti-ZBTB16 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Fornecedor: United States Biological
USBIZ0744-35-FITCEA 852 EUR
USBIZ0744-35-FITC
Anti-ZBTB16 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpos
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!
Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human