Print…

Anti-ZBTB16 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fornecedor: United States Biological
USBIZ0744-35-APEA 852 EUR
USBIZ0744-35-AP
Anti-ZBTB16 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000 Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR