Print…

Anti-ZNF350 Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBIZ0744-26AEA 526 EUR
USBIZ0744-26A
Anti-ZNF350 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ZNF350 (zinc finger protein 350)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR