Print…

Anti-T7 Chicken Polyclonal Antibody

Fornecedor: United States Biological
USBIT0230-03EA 365 EUR
USBIT0230-03
Anti-T7 Chicken Polyclonal Antibody
Recommended dilution: ELISA: 1:60000; Western Blot: 1:5000

Type: Tag
Antigen: T7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR