Print…

Anti-Snai-1a, 1b Rabbit polyclonal antibody

USBIS1014-67L6EA 485 EUR
USBIS1014-67L6
Anti-Snai-1a, 1b Rabbit polyclonal antibody
Recommended dilution: Western Blot: 1:500-1:1000; ELISA: 1:5000-1:20000

Type: Primary
Antigen: Snai-1a, 1b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR