Print…
Anti-SMURF2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Fornecedor: United States Biological
USBIS1014-65E-APEA 852 EUR
USBIS1014-65E-AP
Anti-SMURF2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse