Print…

Anti-SMURF2 Rabbit Polyclonal Antibody

Fornecedor: United States Biological
USBIS1014-65EEA 587 EUR
USBIS1014-65E
Anti-SMURF2 Rabbit Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: SMURF2 (SMAD specific E3 ubiquitin protein ligase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR