Print…

Anti-SMUG1 Goat Polyclonal Antibody

Fornecedor: United States Biological
USBIS1014-65D2EA 633 EUR
USBIS1014-65D2
Anti-SMUG1 Goat Polyclonal Antibody
Anticorpos
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-2?g/ml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: SMUG1 (single-strand-selective monofunctional uracil-DNA glycosylase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR